Kimble™ KIMAX™-35 300mL BOD Bottles
Kimble™ KIMAX™-35 300mL BOD Bottles
Fisher Scientific™

Kimble™ KIMAX™-35 300mL BOD Bottles

For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. SequenceStopper
02885A
also known as FISHERMIS-02885A-12/PK
UnnumberedWith stopper
02885B
also known as FISHERMIS-02885B-EACH
1 to 24With stopper
02885C
also known as FISHERMIS-02885C-EACH
25 to 48With stopper
Catalog number 02885A
also known as FISHERMIS-02885A-12/PK
Price (KRW)
-
No. Sequence:
Unnumbered
Stopper:
With stopper
For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis

Flared mouth forms a water seal to prevent air from being drawn into the bottle during incubation, while shoulder radius provides an interior shape that sweeps entrapped air out of the stopper opening.

  • Type 1, Class B borosilicate glass
  • Available numbered and unnumbered With glass stopper unless otherwise noted
  • Optional snap lid secures stopper in bottle and reduces evaporation of water seal during incubation (sold separately, 02-939)

Specifications
Certifications/ComplianceASTM E438 type I, class A
Diameter (English)2.7 in.
Diameter (Metric)6.9 cm
For Use With (Application)For sampling and incubation of aqueous samples
Height (English)6.5 in.
Height (Metric)16.5 cm
MaterialBorosilicate Glass
No. SequenceUnnumbered
StopperWith stopper
Volume (English)10.14 oz.
Volume (Metric)300 mL
Unit SizePack of 12