PYREX™ Large-Capacity Bottles with Tooled Neck
Fisher Scientific™

PYREX™ Large-Capacity Bottles with Tooled Neck

For distilled water and solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
028873
also known as FISHERMIS-028873-EACH
13,200 mLYes
02887C
also known as FISHERMIS-02887C-EACH
19,000 mLNo
Catalog number 028873
also known as FISHERMIS-028873-EACH
Price (KRW)
-
Capacity (Metric):
13,200 mL
Graduations:
Yes
For distilled water and solutions
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications
Capacity448 oz., 13200 mL
Diameter (English)9.37 in.
Diameter (Metric)238 mm
GraduationsYes
Height (English)17.3 in.
Height (Metric)440 mm
No. per Case4/Cs.
No. per Pack4/Pk.
Through the Closure DispensingYes
Width (English)9.37 in. (Diameter)
Capacity (English)448 oz.
Capacity (Metric)13,200 mL
Closure IncludedNo
ColorClear
GraduatedYes
MaterialBorosilicate Glass
MouthNarrow
Product TypeBottle with Tooled Neck
ShapeCylindrical
SterilityNon-sterile
Unit SizeEach