Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Fisher Scientific™

Easy-Squeeze Wash Bottles, 6/Cs.

Easy to fill and ready to use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorChemical Label
0289712
also known as FIS-0289712
AssortedAssorted Solvents
0289713AssortedAssorted Cleaning
028973BlueDeionized Water
028979BlueSoap
0289711
also known as FIS-0289711
BlueWater
028974BlueDistilled Water
028977
also known as FIS-028977
GreenMethanol
028978NaturalSaline
0289710NaturalUniversal
028972NaturalSodium Hypochlorite (Bleach)
028975NaturalEthanol
028971
also known as FIS-028971
RedAcetone
028976YellowIsopropanol
Catalog number 0289712
also known as FIS-0289712
Price (KRW)
-
Color:
Assorted
Chemical Label:
Assorted Solvents
Easy to fill and ready to use

  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping

Label information includes:

  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement

Not recommended for use above 60°C (140°F).

Specifications
AutoclavableNot Autoclavable
Capacity17 oz., 500mL
Capacity (English)17 oz.
Certifications/ComplianceISO 13485:2003
Closure MaterialPolypropylene
Closure Size38-415 mm
Closure TypePolypropylene Closure
Diameter (English)2.99 in.
Diameter (Metric)76 mm
Height (English)6.81 in.
Height (Metric)173 mm
Product TypeWash Bottle
ShapeCylindrical
Through the Closure DispensingNo
Capacity (Metric)500 mL
Chemical LabelAssorted Solvents
Closure IncludedYes
ColorAssorted
GraduatedNo
MaterialLDPE
MouthWide
Unit SizeCase of 6