Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Fisher Scientific™

Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps

Ideal for soil sampling and environmental applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
02911413
also known as FISHERMIS-02911413-EACH
250 mL70-400 mm
02911412
also known as FISHERMIS-02911412-EACH
125 mL58-400 mm
Catalog number 02911413
also known as FISHERMIS-02911413-EACH
Price (KRW)
-
Capacity (Metric):
250 mL
Closure Size:
70-400 mm
Ideal for soil sampling and environmental applications

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Walls allow for complete removal of contents
  • Wide-mouth design is great for large solid samples
  • Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage

Specifications
Closure ColorWhite
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size70-400 mm
Closure TypeScrew Cap
Diameter (English)2.87 in.
Diameter (Metric)73 mm
Height (English)3.4 in.
Height (Metric)88 mm
Through the Closure DispensingYes
Capacity (English)8 oz.
Capacity (Metric)250 mL
Certifications/ComplianceUSP Type III
Closure IncludedYes
ColorClear
MaterialSoda Lime Glass
MouthWide
Product TypeStraight-sided Jar
ShapeRound
Unit SizeEach