Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap
Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap
Fisher Scientific™

Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck FinishQuantity
02911456A
also known as FIS-02911456A
240 mL58-40024/Cs.
02911465A
also known as FIS-02911465A
30 mL33-400432/Cs.
02911453A
also known as FIS-02911453A
30 mL33-40048/Cs.
02911466A
also known as FIS-02911466A
60 mL38-400288/Cs.
02911467A
also known as FIS-02911467A
120 mL48-400144/Cs.
02911468A240 mL58-40096/Cs.
02911469A480 mL70-40048/Cs.
Catalog number 02911456A
also known as FIS-02911456A
Price (KRW)
-
Capacity (Metric):
240 mL
Neck Finish:
58-400
Quantity:
24/Cs.
Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.
  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents

White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
AutoclavableAutoclavable
Closure ColorWhite
Closure LinerWith Liner
Closure MaterialPolypropylene
Closure Size58-400 mm
Closure TypeScrew Cap
Diameter (Metric)61 mm
Liner MaterialPulp/Vinyl
Neck Finish58-400
Quantity24/Cs.
Through the Closure DispensingYes
Capacity (English)8 oz.
Capacity (Metric)240 mL
Closure IncludedYes
ColorClear
GraduatedYes
MaterialGlass
MouthWide
Product TypeGraduated Medium Round Bottle
ShapeRound
Unit SizeCase of 24