Natural HDPE Wide Mouth Oblong Bottles with White Polypropylene PE Foam Cap
Natural HDPE Wide Mouth Oblong Bottles with White Polypropylene PE Foam Cap
Fisher Scientific™
Natural HDPE Wide Mouth Oblong Bottles with White Polypropylene PE Foam Cap
Ideal for sampling solids and powders. Fisherbrand™ White HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap offer a space saving design for the workbench.
Have Questions?
Catalog NumberCapacity (Metric)
02911950A1250 mL
Catalog number 02911950A
Price (KRW)
-
Capacity (Metric):
1250 mL
Natural HDPE Wide Mouth Oblong Bottles offer a space saving design for the workbench. These bottles feature a wide mouth opening, ideal for sampling solids and powders. HDPE provides an excellent moisture barrier and are impact resistant. They are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.
  • HDPE provides moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
  • Polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Specifications
AutoclavableNot Autoclavable
Closure ColorWhite
Closure LinerWith Liner
Closure MaterialPolypropylene
Closure Size53-400 mm
Closure TypeScrew Cap
Liner MaterialPE Foam
Neck Finish53-400
Quantity6/Cs.
Through the Closure DispensingNo
Capacity (Metric)1250 mL
Closure IncludedYes
ColorWhite, Natural
GraduatedNo
MaterialHDPE
MouthWide
Product TypeOblong Bottle
ShapeOblong
Unit SizeCase of 6