Natural Polypropylene Jars with White Polypropylene Unlined Cap
Natural Polypropylene Jars with White Polypropylene Unlined Cap
Fisher Scientific™

Natural Polypropylene Jars with White Polypropylene Unlined Cap

Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap are translucent and durable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)QuantityShape
02912027A960 mL24/Cs.Round
02912025A30 mL72/Cs.Round
02912026A60 mL48/Cs.Round
02912028A120 mL36/Cs.Round
02912029A240 mL36/Cs.Round
02912024A480 mL24/Cs.Straight-sided, Round
Catalog number 02912027A
Price (KRW)
-
Capacity (Metric):
960 mL
Quantity:
24/Cs.
Shape:
Round
Natural Polypropylene Jars are translucent and durable. Jars are ideal for storing and collecting specimens and lab supplies. Wide neck and straight sides facilitate easy recovery of contents. Polypropylene offers greater tolerance to high temperatures than HDPE and better contact clarity.
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Polypropylene material offers greater tolerance to high temperatures than HDPE and better contact clarity
Specifications
ClarityTranslucent
Closure ColorWhite
Closure LinerUnlined
Closure MaterialPolypropylene
Closure TypeScrew Cap
For Use With (Application)Storing and Collecting Specimens and Lab Supplies
IncludesCap
Liner MaterialUnlined
Neck Finish120-400
Quantity24/Cs.
Capacity (English)32 oz.
Capacity (Metric)960 mL
ColorNatural
MaterialPolypropylene
MouthWide
Product TypeJar
ShapeRound
Unit SizeCase of 24