Black Phenolic PolyCone Cap
Fisher Scientific™
Black Phenolic PolyCone Cap
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck FinishQuantity
0291205622-400576/Cs.
0291180320-4005500/Cs.
0291205524-400576/Cs.
02911806A
also known as FIS-02911806A
24-4004100/Cs.
0291210128-400576/Cs.
02912091A
also known as FIS-02912091A
33-4002200/Cs.
0291206338-400576/Cs.
Catalog number 02912056
Price (KRW)
-
Neck Finish:
22-400
Quantity:
576/Cs.
PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
Specifications
Liner MaterialPolyCone
Neck Finish22-400
Quantity576/Cs.
ColorBlack
MaterialPhenolic Resin
Product TypePolycone Cap
Unit SizePack of 12