Search
| Catalog Number | Neck Finish | Quantity |
|---|---|---|
| 02912080 | 89-400 | 144/Cs. |
| 02912066 | 18-400 | 100/Pk. |
| 02912067 | 20-400 | 100/Pk. |
| 02912071 | 33-400 | 100/Pk. |
| 02912072 | 38-400 | 500/Cs. |
| 02912073 | 43-400 | 500/Cs. |
| 02912074 | 45-400 | 500/Cs. |
| 02912075 | 48-400 | 100/Pk. |
| 02912076 | 53-400 | 100/Pk. |
| 02912077 | 58-400 | 100/Pk. |
| 02912078 | 70-400 | 144/Cs. |
| 02912079 | 83-400 | 144/Cs. |
Polypropylene (PP) caps with F217 & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps. They have a PTFE facing, backed by F217 liners. The F217 liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE). PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference.