White Polypropylene Pulp/Vinyl Cap
Fisher Scientific™
White Polypropylene Pulp/Vinyl Cap
Commonly used for food, beverage, medical and chemical applications packed at less than 120°F. Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner are known for good impact strength and resistant to mild acids, alkalis, solvents, alcohols, oils and aqueous products.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck FinishQuantity
02912291A48-400576 Each/Cs.
02911853A
also known as FIS-02911853A
70-400760/Cs.
02912292A
also known as FIS-02912292A
53-400288 Each/Cs.
02912088A
also known as FIS-02912088A
70-400144 Each/Cs.
Catalog number 02912291A
Price (KRW)
-
Neck Finish:
48-400
Quantity:
576 Each/Cs.
Pulp/Vinyl Lined Polypropylene Caps are commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products. Polypropylene caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability.
Polypropylene caps are:
  • Known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Autoclavable
Pulp/Vinyl liners are:
  • Made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
  • Commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
AutoclavableAutoclavable
Liner MaterialPulp/Vinyl
Neck Finish48-400
PackagingPacked in 48 bags of 12
Quantity576 Each/Cs.
ColorWhite
MaterialPolypropylene
Product TypeLined Screw Cap
ShapeRound
Unit SizeCase of 576