You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Vials
▸
Scintillation Vials and Caps
▸
20mL Borosilicate Glass Scintillation Vials with White Urea Caps
Fisher Scientific™
20mL Borosilicate Glass Scintillation Vials with White Urea Caps
Fit all common counters
Have Questions?
Change view
Catalog Number
Material (Liner)
Closure Type
033375
also known as KIMBLE-033375-500/CS
Foil/Cork
Attached Screw Cap
033377
also known as FIS-033377
Polyethylene
Separate Screw Cap
033404E
Foil/Cork
Separate Screw Cap
3 Options
Catalog number
033375
also known as KIMBLE-033375-500/CS
Price (KRW)
-
Material (Liner):
Foil/Cork
Closure Type:
Attached Screw Cap
Fit all common counters
Potassium-free glass provides consistently low activity counts and excellent light transmission
Caps fit GPI 22-400 thread finish and are suitable for marking
Specifications
Closure Material
Urea
Closure Size
22-400
Diameter (English) Outer
1.12 in.
Height (English)
2.37 in.
Height (Metric)
61 mm
Material (Liner)
Foil/Cork
Packaging Type
Shrink-wrapped Tray
Product Type
Scintillation Vial
Volume (English)
0.67 oz.
Volume (Metric) Total
20 mL
Closure Type
Attached Screw Cap
Color
White
Diameter (Metric) Outer
28 mm
Marking Spot
Yes
Material
Borosilicate Glass
Volume (Metric)
20 mL
Unit Size
Case of 500