Specifications
Certifications/ComplianceUSP type I, ASTM E438
GraduatedYes
Graduations0.2 mL
Negative Graduations5 mL
Tolerance±1 mL
ColorGreen
DisposableSingle-use
MaterialBorosilicate Glass
SterilitySterile
TypeSerological Pipet
Volume (Metric)25 mL
Unit SizeCase of 200