Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops
Fisher Scientific™
Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops
Open top closures providing excellent chemical resistance
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck Finish
033407G18-400
033407E
also known as FIS-033407E
15-425
Catalog number 033407G
Price (KRW)
-
Neck Finish:
18-400
Open top closures providing excellent chemical resistance
  • Phenolic
  • Open top
  • Autoclavable
  • Unlined
  • Black
Specifications
AutoclavableAutoclavable
For Use With (Equipment)Bottle
Neck Finish18-400
No. per Case144 Cs.
ColorBlack
MaterialPhenolic Resin
Product TypeOpen Top Screw Cap
Unit SizeCase of 144