11 mm Autosampler Vial Crimp Caps with Septa, Aluminum Seal, 5.5 mm Hole
Fisher Scientific™
11 mm Autosampler Vial Crimp Caps with Septa, Aluminum Seal, 5.5 mm Hole
Fisherbrand™'s 11 mm crimp caps with pre-assembled septa are convenient, minimize contamination from handling and are compatible with most HPLC and GC autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSeptumColor (Seal)
033913PTFE/RubberClear/Red
0339133PTFE/SiliconeRed/White
0339134PTFE/Silicone/PTFERed/White/Red
Catalog number 033913
Price (KRW)
-
Septum:
PTFE/Rubber
Color (Seal):
Clear/Red
Fisherbrand™'s 11 mm crimp caps with pre-assembled septa are convenient, minimize contamination from handling and are compatible with most HPLC and GC autosamplers.
  • Aluminum crimp closures provide a secure leak-resistant seal
  • Aluminum seals must be applied with a crimping tool
  • Synthetic PTFE/Red Rubber seal is specially formulated for improved background performance
Specifications
Closure Size11 mm
Color (Seal)Clear/Red
Diameter (Metric) Hole5.5 mm
Hardness60 Shore A
No. per Pack100
Product TypeCrimp Cap
SeptumPTFE/Rubber
Thickness (Metric)1 mm
ColorSilver
MaterialAluminum
Pre-slitNo
Product LineFisherbrand
Unit SizePack of 100