Specifications
Base StyleConical with Polyspring
Certifications/ComplianceCertificate of Conformance (COC)
DisposableSingle-use
For Use With (Application)Micro Sampling
For Use With (Equipment)Wide Opening Vial
Height (Metric)29 mm
Insert TypeInserts with spring for HPLC and GC Vials
No. per Pack100
Packaging Type10 Packs of 100
Product TypePolyspring Insert
Volume (Metric) Residual1 μL
Volume (Metric) Total0.375 mL
Volume (Metric) Usable0.3 mL
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass, Polypropylene
Product LineFisherbrand
Quantity100
SterilityNon-sterile
Volume (Metric)300 μL
Unit SizePack of 100