Solvent-Vented™ and Safety-Labeled Wash Bottles
Fisher Scientific™

Solvent-Vented™ and Safety-Labeled Wash Bottles

Are pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Label
0340930Ethyl Acetate
0340937
also known as FISHERMIS-0340937-3/PK
Acetone
0340931Methyl Ethyl Ketone
Catalog number 0340930
Price (KRW)
75,000
Pack of 3
Chemical Label:
Ethyl Acetate
Price (KRW)
75,000
Pack of 3
Are pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.
  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

 

Use LYOB only with solvents compatible with LDPE.


Specifications
AutoclavableNot Autoclavable
Capacity16 oz., 500mL
Closure ColorGreen
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypePolypropylene Closure
Through the Closure DispensingYes
Capacity (English)16 oz.
Capacity (Metric)500 mL
Certifications/ComplianceOSHA 29 CFR 1910.1200
Chemical LabelEthyl Acetate
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialLDPE
MouthWide
Product TypeWash Bottle
ShapeCylindrical
Unit SizePack of 3