Search
| Catalog Number | Color |
|---|---|
| 0344816 | Fluorescent Orange |
| 0344812 also known as FIS-0344812 | Blue |
| 0344811 | Natural |
| 0344813 | Fluorescent Green |
| 0344814 | Fluorescent Pink |
| 0344815 | Fluorescent Yellow |
| 0344817 also known as FIS-0344817 | Assorted |
Customize your tube rack. Fisherbrand™ Interlocking Four-Way Tube Racks employ an ingenious system of interlocking, 4-way racks. Rotate racks to accommodate 50mL, 15mL, 1.5mL or 0.5mL tubes, then link them together for whatever size rack you need. Made of sturdy, autoclavable polypropylene.
Create a customized tube rack: rotate and connect.
Rotate each rack for the tube size you need. Each rack can accommodate:
Connect racks to increase the capacity of your tube rack: