White Nylon Bristle Brush
Fisher Scientific™

White Nylon Bristle Brush

Fisherbrand™ White Nylon Bristle Brushes are heavy-duty brushes for cleaning beakers, jars, and similar containers.
Have Questions?
Catalog NumberColor
03536
also known as FIS-03536
White
Catalog number 03536
also known as FIS-03536
Price (KRW)
-
Color:
White
Fisherbrand™ White Nylon Bristle Brushes are heavy-duty brushes for cleaning beakers, jars, and similar containers.

  • Extra-heavy bristle with double-tufted end
  • Wooden handle
  • Chemical-resistant

Specifications
ColorWhite
Diameter (English) Bristle2.87 in.
Diameter (Metric) Bristle7.2 cm
For Use With (Application)For cleaning beakers, jars, and similar containers
Length (English)16 in.
Length (English) Bristle5 in.
Length (Metric)40.64 cm
Length (Metric) Bristle12.7 cm
SizeMedium
Handle MaterialWood
Product TypeBristle Brush
Tip TypeDouble Tufted End
Unit SizeEach