You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Brushes
▸
Babcock Cream Bottles Brush
Fisher Scientific™
Babcock Cream Bottles Brush
Fan-style tip for cleaning Babcock bottles.
Have Questions?
Catalog Number
Color
03640
also known as FIS-03640
Black
Catalog number
03640
also known as FIS-03640
Price (KRW)
-
Color:
Black
Fan-style tip for cleaning Babcock bottles.
Bristle portion on tip, 1.75 dia. × 2 in.L (4.4 × 5cm)
Upper portion, 0.5 dia. × 3 in.L (1.3 × 7.6cm)
Specifications
Color
Black
For Use With (Application)
For cleaning babcock bottles
Includes
Metal ferrule
Length (English)
11.5 in.
Length (Metric)
29.2 cm
Size
Small
Product Type
Babcock Brush
Tip Type
Fan Tip
Unit Size
Each