SMART 3™ Colorimeter Water Quality Analyzer
Fisher Scientific™

SMART 3™ Colorimeter Water Quality Analyzer

Multitest colorimeter measures numerous water quality parameters with laboratory accuracy and the convenience of a portable, handheld system
Have Questions?
Catalog NumberWavelength Range
03920552
also known as FISHERMIS-03920552-12/CS
428, 520, 568, 635 nm
Catalog number 03920552
also known as FISHERMIS-03920552-12/CS
Price (KRW)
-
Wavelength Range:
428, 520, 568, 635 nm
Multitest colorimeter measures numerous water quality parameters with laboratory accuracy and the convenience of a portable, handheld system

Simple menu-driven operation displays results in concentration (ppm) or absorbance.

  • Waterproof to IP67
  • More than 75 preprogrammed tests in library and 25 user-entered tests
  • Choose any of four wavelengths (see Specifications) to determine absorbance of a sample at a desired wavelength—proper wavelength selection is assured, since all optical filters are included and automatically selected
  • Autoblank and auto-off
  • Using optional SMARTLink 3 Program and cable (order online), the SMART 3 Colorimeter can interface with any DOS-based computer program to send test data to a database or ASCII delimited file

Specifications
Accuracy±0.005 AU at 1 AU
Certifications/ComplianceCE, EPA
ConnectionsMini B USB
Dimensions (L x W x H)3.5 x 7.5 x 2.5 in. (8.84 x 19.05 x 6.35 cm)
For Use With (Application)Industrial water boiling and cooling, water and wastewater
Height (English)2.5 in.
Height (Metric)6.35 cm
IncludesLithium ion rechargeable battery and USB wall charger, USB cable, four sample tubes, instrument manual including test procedures and quick start guide
Length (English)3.5 in.
Length (Metric)8.84 cm
Pathlength10 mm
ResolutionDetermined by reagent system
Warranty2 Years
Weight (English)13 oz.
Weight (Metric)362 g
Width (English)7.5 in.
Width (Metric)19.05 cm
DisplayBacklit LCD
IP RatingIP67
LanguagesSeven user-selectable languages
Light SourceLED
Voltage3.7 V
Wavelength Range428, 520, 568, 635 nm
Unit SizeCase of 12