RO-TAP™ II Sieve Shaker Maintenance Kits
RO-TAP™ II Sieve Shaker Maintenance Kits
Fisher Scientific™
RO-TAP™ II Sieve Shaker Maintenance Kits
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
0490914
also known as FISHERTOPSELLING-0490914-EACH
RX-29/30 Ro-Tap Sieve Shaker
0490913
also known as FISHERTOPSELLING-0490913-EACH
RX-29/30/94 Ro-Tap Sieve Shaker
0490915
also known as FISHERTOPSELLING-0490915-EACH
RX-94 Ro-Tap sieve shakers
Catalog number 0490914
also known as FISHERTOPSELLING-0490914-EACH
Price (KRW)
-
For Use With (Equipment):
RX-29/30 Ro-Tap Sieve Shaker
For replacement of parts due to normal wear and tear.
Specifications
For Use With (Equipment)RX-29/30 Ro-Tap Sieve Shaker
IncludesTiming belt, cam gear, timing belt pulley
Product TypeSieve Shaker Maintenance Kit
Unit SizeEach