You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Tubes
▸
Microtubes
▸
Premium Microcentrifuge Tubes: 0.6mL
Fisher Scientific™
Premium Microcentrifuge Tubes: 0.6mL
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Have Questions?
Change view
Catalog Number
Color
05408124
Green
05408121
Red
05408123
Blue
05408120
also known as FIS-05408120
Natural
05408122
Yellow
05408126
Orange
05408125
Amber
05408127
Lavender
05408128
Assorted
9 Options
Catalog number
05408124
Price (KRW)
-
Color:
Green
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Capacity: 0.6mL
High-clarity polypropylene
Graduated in 0.1mL markings
Withstand forces up to 30,000 × g
Easy-open caps with needle insertion spot and textured marking area
Labeling area on tube exterior
Highly polished interior for maximum sample recovery
Temperature range of -80° to +121°C
Fit all standard rotors
Nonpyrogenic
Autoclavable at 121°C/15psi for 15 minutes
Specifications
Cap Style
Snap Cap with Flat Top
Certifications/Compliance
RNase, DNase-free
Diameter (Metric)
7.5 mm
Includes
Needle insertion spot
Label Type
Labeling Area
Length (Metric)
30.5 mm
Autoclavable
Autoclavable
Capacity (Metric)
0.5 mL (0.6 mL max.)
Closure Type
Flat Snap Cap
Color
Green
Graduated
Yes
Material
Polypropylene
Max. RCF
30,000 x g
Unit Size
Pack of 500