You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Pipette Products
▸
Pipette Tips
▸
Universal Pipette Tips
▸
Premium Plus MultiFlex Gel-Loading Tips
Fisher Scientific™
Premium Plus MultiFlex Gel-Loading Tips
Ideal for gel electrophoresis
Have Questions?
Change view
Catalog Number
Packaging
Shape
05408150
also known as FISHERMIS-05408150-EACH
400/cs.
Flat
05408154
also known as FISHERTOPSELLING-05408154-5/PK
1000/bag
Round
05408151
400/cs.
Round
3 Options
Catalog number
05408150
also known as FISHERMIS-05408150-EACH
Price (KRW)
-
Packaging:
400/cs.
Shape:
Flat
Ideal for gel electrophoresis
Easily access bottom of narrow vessels and gel wells
Universal fit
Graduated marks for quick volume checks
Certified DNase-, RNase-, and human DNA-free
Match O.D. to Gel Thickness
Round 0.57mm O.D. tips are recommended for loading gels ≥0.6mm thick—great for SDS Page and 2-D gels
Flat tips are recommended for gels up to 0.4mm thick
Use flat 0.37mm O.D. tips for 0.4mm thick PAGE sequencing gels; flat 0.17mm tips for 0.2mm DNA sequencing gels
MultiFlex Tips
0.5 to 200μL volume
Fit research-grade pipetters
83mm long
5μL graduation
Specifications
For Use With (Application)
Gel electrophoresis
Graduated
Yes
Graduations
5μL
Length (English)
3.26 in.
Length (Metric)
8.3cm
Packaging
400/cs.
Shape
Flat
Thickness
0.4mm
For Use With (Equipment)
Universal Fit
Tip Style
Gel Loading
Volume (Metric)
0.5 to 200 μL
Unit Size
Each