You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Kimble™ Black Phenolic Screw-Thread Closures
DWK Life Sciences
Kimble™ Black Phenolic Screw-Thread Closures
Highly resistant to temperature and steam during autoclaving
Have Questions?
Catalog Number
Neck Finish
055695TP
also known as FISHERMIS-055695-EACH
15-415
Catalog number
055695TP
also known as FISHERMIS-055695-EACH
Price (KRW)
360,000
Each
Add to cart
Neck Finish:
15-415
Price (KRW)
360,000
Each
Add to cart
Cemented-in PTFE-faced 14B white rubber liner
Three screw-thread finishes
Specifications
Autoclavable
Autoclavable
For Use With (Equipment)
Bottle
Liner Material
PTFE-faced Rubber
Neck Finish
15-415
Closure Included
Yes
Color
Black
Material
Phenolic Resin
Product Type
Screw Cap
Unit Size
Each