You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Stoppers and Closures
▸
Bottle Caps
▸
Wheaton™ Microlink Caps
Fisher Scientific™
Wheaton™ Microlink Caps
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
Have Questions?
Catalog Number
Neck Finish
Color
Product Type
No. per Case
0645012
also known as FISHERTOPSELLING-0645012-EACH
33-430
Black
Open Top Screw Cap
50/Cs.
Catalog number
0645012
also known as FISHERTOPSELLING-0645012-EACH
Price (KRW)
-
Neck Finish:
33-430
Color:
Black
Product Type:
Open Top Screw Cap
No. per Case:
50/Cs.
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
Autoclavable
Specifications
Autoclavable
Autoclavable
For Use With (Equipment)
Bottle
Liner Material
Polypropylene/Silicone/PTFE
Neck Finish
33-430
No. per Case
50/Cs.
Certifications/Compliance
ISO 9001
Color
Black
Material
Polypropylene
Product Type
Open Top Screw Cap
Unit Size
Each