Specifications
Accuracy±0.01%
Certifications/ComplianceNIST, ISO 17025
ColorWhite
For Use With (Application)For general purpose timing applications
Height (English) Display1 in.
Height (Metric) Display2.5 cm
IncludesWall mount bracket, stand, AAA battery, Traceable certificate, instructions
AlarmsAudible/Visible
DisplayLCD
Product TypeBenchtop Timer
Time Range24 hr. to 1 sec.
Weight (English)5 oz.
Weight (Metric)141 g
Unit SizeEach