Search
Search

| Catalog Number | Pressure Range | Product Type | Weight (Metric) |
|---|---|---|---|
| 0666421 also known as FISHERTOPSELLING-0666421-EACH | -30 to 30 psi | Non-Printing Model | 0.14 kg |
| 0666419 also known as FISHERTOPSELLING-0666419-EACH | -15 to 15 psi | Non-Printing Model | 0.14 kg |
| 0666418 also known as FISHERTOPSELLING-0666418-EACH | -5 to 5 psi | Non-Printing Model | 0.14 kg |
| 0666422 also known as FISHERTOPSELLING-0666422-PK | -100 to 100 psi | Non-Printing Model | 0.14 kg |
Portable units measure and display differential pressure/vacuum in 11 different units (psi, mbar etc)
Printing Model