Specifications
Cap StyleSnap Cap
Certifications/ComplianceRNase, DNase-free
Closure ColorNatural
Closure MaterialPolypropylene
No. per Pack250/Pk.
Product TypeMicrocentrifuge Tube
AutoclavableAutoclavable
Bottom ShapeConical
Capacity (Metric)1.7 mL
Closure TypeSnap Cap
ColorNatural
GraduatedYes
MaterialPolypropylene
Max. RCF17,000 x g
SterilityNon-sterile
Unit SizeCase of 40