Cell Scrapers
Fisher Scientific™
Cell Scrapers
Angled swivel blade minimizes cell damage and ensures even contact with growth area. Corning™ Cell Scrapers are ideal for harvesting cells.
Have Questions?
Catalog NumberLength (Metric) HandleLength (Metric) Blade
07200365
also known as FISHERMIS-07200365-EACH
25 cm1.8 cm
Catalog number 07200365
also known as FISHERMIS-07200365-EACH
Price (KRW)
-
Length (Metric) Handle:
25 cm
Length (Metric) Blade:
1.8 cm
Angled swivel blade minimizes cell damage and ensures even contact with growth area. Corning™ Cell Scrapers are ideal for harvesting cells.
  • Long handle permits easy access to cell culture flasks and roller bottles 
  • Designed for use in 25, 75 and 225cmflasks
  • Sterile; certified nonpyrogenic
  • Individually wrapped
Specifications
For Use With (Application)Harvesting Cells
IncludesBlade with handle
Length (English) Handle10 in.
Length (Metric) Handle25 cm
SterilitySterile
Length (English) Blade0.7 in.
Length (Metric) Blade1.8 cm
Product TypeCell Scraper
Unit SizeEach