Drierite™ Drying Desiccants, W.A. Hammond
Drierite™ Drying Desiccants, W.A. Hammond
Drierite™ Drying Desiccants, W.A. Hammond
Fisher Scientific™

Drierite™ Drying Desiccants, W.A. Hammond

Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 lb.
5 lb.
Mesh Size:
8 Mesh
Catalog number 075773A
also known as FISHERTOPSELLING-075773A-EACH
Price (KRW)
-
Quantity:
1 lb.
Mesh Size:
8 Mesh
Request bulk or custom format
Specifications
CAS7778-18-9
ChEBICHEBI:31346
Chemical Name or MaterialCalcium Sulfate
IUPAC Namecalcium sulfate
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
View more
Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
  • All purpose desiccant made of anhydrous calcium sulfate
  • Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
  • Dries organic liquids to low ppm levels
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas dryers, glove boxes, sealed chambers
  • Useful for water removal from liquid reaction mixtures and multi-component gas streams.
  • Neutral, stable, constant in volume, and inert except toward water
  • Insoluble in organic solvents
  • Absorbs moisture at temperatures up to 66°C