Cell Scrapers
Cell Scrapers
Cell Scrapers
Cell Scrapers
Fisher Scientific™
Cell Scrapers
Easily harvest cells from tissue culture flasks
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English) HandleLength (English) BladeSize
08100242
also known as FIS-08100242
15 in.1.18 in.Large
08100241
also known as FIS-08100241
10 in.0.7 in.Small
Catalog number 08100242
also known as FIS-08100242
Price (KRW)
-
Length (English) Handle:
15 in.
Length (English) Blade:
1.18 in.
Size:
Large
Easily harvest cells from tissue culture flasks
Fisherbrand Cell Scrapers are appropriate for a variety of cell culture vessels. The uniquely designed handle and blade reduce cell damage during the harvesting process.
  • Presterilized and individually wrapped in peel-open packaging
  • Nonpyrogenic
  • Polyethylene copolymer blade pivots to permit even contact with growth surface
  • Ribbed polystyrene handle provides secure grip and added control
  • Flexible joint between blade and handle eases entry into vessel
  • Pivoting blade improves position control to adjust for difficult angles during cell harvesting
Specifications
Blade MaterialPolyethylene Copolymer
For Use With (Equipment)For a variety of cell culture vessels
Length (English) Handle15 in.
Length (Metric) Handle39 cm
SizeLarge
Length (English) Blade1.18 in.
Length (Metric) Blade3 cm
MaterialPolystyrene (Handle)
Product TypeCell Scraper
Unit SizeCase of 100