Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device
Fisher Scientific™

Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device

Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCOClosure ColorHeight (Metric)
08607079
also known as FIS-08607079
0.1 to 0.5 kDaGreen63 mm
08607075
also known as FIS-08607075
8 to 10 kDaYellow44 mm
08607077
also known as FIS-08607077
50 kDaWhite44 mm
08607080
also known as FIS-08607080
0.5 to 1 kDaOrange63 mm
08607082
also known as FIS-08607082
8 to 10 kDaYellow63 mm
08607085
also known as FIS-08607085
100 kDaBlue63 mm
Catalog number 08607079
also known as FIS-08607079
Price (KRW)
-
MWCO:
0.1 to 0.5 kDa
Closure Color:
Green
Height (Metric):
63 mm
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever

Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.

  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume

Specifications
Capacity (Metric)400 to 500 μL
Closure ColorGreen
Closure MaterialPolypropylene
Closure TypeLuer lok
Diameter (English) Membrane0.25 in.
Diameter (Metric) Membrane6.4 mm
For Use With (Application)Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
Height (English)2.4 in.
Height (Metric)63 mm
Kit Contents1mL syringe
MaterialPolycarbonate
MembraneCellulose Ester
Product TypeMicro Float-A-Lyzer Dialysis Device
QuantityEach
Width (English)1.77 in.
Width (Metric)45 mm
MWCO0.1 to 0.5 kDa
Unit SizePack of 12