Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Fisher Scientific™

Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO

Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)MWCO
0867015D
also known as FISHERTOPSELLING-0867015D-EACH
31.8 mm8 kDa
0867012B
also known as FISHERMIS-0867012B-EACH
11.5 mm1 kDa
0867012C
also known as FISHERTOPSELLING-0867012C-EACH
24 mm1 kDa
0867012D
also known as FISHERMIS-0867012D-EACH
29 mm1 kDa
0867013C
also known as FISHERTOPSELLING-0867013C-EACH
24 mm2 kDa
0867013D
also known as FISHERTOPSELLING-0867013D-EACH
29 mm2 kDa
0867014C
also known as FISHERMIS-0867014C-EACH
34.4 mm3.5 kDa
0867016D
also known as FISHERTOPSELLING-0867016D-EACH
29 mm10 kDa
0867019C
also known as FISHERTOPSELLING-0867019C-EACH
21.6 mm50 kDa
0867014B
also known as FISHERTOPSELLING-0867014B-EACH
29 mm3.5 kDa
Catalog number 0867015D
also known as FISHERTOPSELLING-0867015D-EACH
Price (KRW)
-
Diameter (Metric):
31.8 mm
MWCO:
8 kDa
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane

This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.

  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg

Do not tie knots in tubing; use closures

Specifications
Diameter (English)1.25 in.
For Use With (Application)Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
FormulationPackaged wet in 0.05% sodium azide
Length (English)33 ft.
Length (Metric)10 m
MaterialRegenerated Cellulose
Product TypeSpectra/Por 6 Dialysis Tubing
QuantityEach
Temperature Range (English)39.2°F to 249.8°F
Temperature Range (Metric)4°C to 121°C
Diameter (Metric)31.8 mm
MWCO8 kDa
Unit SizeEach