Search
| Catalog Number | Diameter (Metric) | Length (Metric) |
|---|---|---|
| 08670C also known as FISHERTOPSELLING-08670C-EACH | 25.5 mm | 30.5 m |
| 08670A also known as FIS-08670A | 14.6 mm | 30.5 m |
| 08670D also known as FIS-08670D | 32 mm | 30.5 m |
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
Do not tie knots in tubing; use closures