Superfrost™ Disposable Microscope Slides
Fisher Scientific™

Superfrost™ Disposable Microscope Slides

Superfrost coating impervious to most common laboratory chemicals and reagents
Have Questions?
Catalog NumberDescription
12550133
also known as FISHERMIS-12550133-EACH
Superfrost Slide, 1.2mm thick
Catalog number 12550133
also known as FISHERMIS-12550133-EACH
Price (KRW)
-
Description:
Superfrost Slide, 1.2mm thick
Superfrost coating impervious to most common laboratory chemicals and reagents

  • Brilliant white, opaque coating takes and retains most laboratory marking methods
  • Each side has a written key for easy orientation; the word “SPECIMEN” instantly tells you which side is up
  • Slides can be imprinted with your laboratory or hospital's name (call your Fisher HealthCare™ Sales Representative for details)

Specifications
CoatingOpaque
DescriptionSuperfrost Slide, 1.2mm thick
Dimensions (L x W)3 x 1 in. (75 x 25 mm)
Length (English)3 in.
Width (English)1 in.
ColorWhite
FrostedSuperFrost
Length (Metric)76 mm
Thickness (Metric)1.2 mm
TypeDisposable Microscope Slide
Width (Metric)25 mm
Unit SizeEach