accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
accumet™ XL150 Benchtop pH Meters
Fisher Scientific™

accumet™ XL150 Benchtop pH Meters

Provide advanced pH measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
13636XL150
also known as ACCUMET-13636XL150-EACH
pH Meter, NIST Certificate
13636XL150AMeter with pH Kit, NIST Cert.
Catalog number 13636XL150
also known as ACCUMET-13636XL150-EACH
Price (KRW)
-
Type:
pH Meter, NIST Certificate
Provide advanced pH measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Specifications
AccuracyMultiple
Accuracy (Temperature)±0.3°C
Accuracy (mV)±0.2
Accuracy (pH)±0.002
Calibration Points (pH)Up to 6 pre-set or 5 custom
Certifications/ComplianceNIST, ISO, FDA, USDA, EPA
Detection RangeMultiple
Dimensions (L x W x H)24.6 x 17.3 x 9.9 cm (9.7 x 6.8 x 3.9 in.)
Frequency50/60 Hz
Height (English)3.89 in.
Height (Metric)9.9 cm
IncludesMeter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply
InputsBNC, ATC
Length (English)9.68 in.
Length (Metric)24.6 cm
ModelXL150 pH Kit
OutputsRS-232, USB-A, Mini-B USB, RJ45, stirrer port (2)
ResolutionMultiple
Resolution (Temperature)0.1°C
Resolution (mV)0.1 mV
TypepH Meter, NIST Certificate
Voltage100/240 V
Warranty3 Years
Weight (English)2.5 lb.
Weight (Metric)1.13 kg
Width (Metric)17.3 cm
Calibration Points1 to 6
Data Points2000
Display TypeLCD
Range (mV)±2000
Range (pH)-2 to 20
Resolution (pH)0.001, 0.01, 0.1
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach