accumet™ XL200 Benchtop pH/Conductivity Meters
accumet™ XL200 Benchtop pH/Conductivity Meters
accumet™ XL200 Benchtop pH/Conductivity Meters
accumet™ XL200 Benchtop pH/Conductivity Meters
accumet™ XL200 Benchtop pH/Conductivity Meters
Fisher Scientific™

accumet™ XL200 Benchtop pH/Conductivity Meters

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL200 pH/Conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
13636XL200
also known as ACCUMET-13636XL200-EACH
pH/Cond Meter, NIST Cert.
13636XL200AMeter, pH/Cond. Kit, NIST Cert.
Catalog number 13636XL200
also known as ACCUMET-13636XL200-EACH
Price (KRW)
-
Product Type:
pH/Cond Meter, NIST Cert.
Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL200 pH/Conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Display up to three measurements simultaneously (two pH or ISE, one conductivity)
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, 8 Pin DIN (conductivity), ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Specifications
AccuracyMultiple
Accuracy (Conductivity)±1% FS
Accuracy (Resistivity)± 1% full scale
Accuracy (Salinity)± 1% full scale
Accuracy (TDS)± 1% full scale
Accuracy (mV)±0.2 mV
Accuracy (pH)±0.002
Calibration Points (Conductivity)Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Calibration Points (pH)Up to 6 preset or 5 custom
Cell Constant0.01 to 10
Certifications/ComplianceNIST, ISO, FDA, USDA, EPA
Data Management Details3 to 86400 sec.
Data Points2000
Detection RangeMultiple
Dimensions (L x W x H)24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
Frequency50/60Hz
Height (English)3.89 in.
Height (Metric)9.9 cm
IncludesMeter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp. probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply
InputsBNC, ATC
Length (English)9.68 in.
Length (Metric)24.6 cm
ModelXL200 pH/Conductivity Kit
OutputsRS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Product TypepH/Cond Meter, NIST Cert.
ResolutionMultiple
Resolution (Conductivity)0.001/0.01/0.1mS
Resolution (Resistivity)0.01/0.1W, 0.001/0.1kW, 0.01MW
Resolution (Salinity)0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (TDS)0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (mV)0.1mV
Temperature Accuracy±0.2°C
Temperature Resolution0.1°C
Voltage100/240 V
Warranty3 Years
Weight (English)2.5 lb.
Weight (Metric)1.13 kg
Width (Metric)17.3 cm
Display TypeLCD
Measurement ModespH, mV, Conductivity, TDS, Salinity, Resistivity, Temperature
Range (Conductivity)0 to 500 mS
Range (Resistivity)3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW
Range (Salinity)0 to 80 ppt
Range (TDS)0.050 ppm to 500 ppt (at TDS factor 1.00)
Range (mV)±2000 mV
Range (pH)-2 to 20
Resolution (ISE)0.001/0.01/0.1/1 (Automatic)
Resolution (pH)0.1/0.01/0.001
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach