accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
Fisher Scientific™

accumet™ XL250 Benchtop pH/ISE Dual Channel Meters

Provide advanced pH and ion measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL250 pH/ISE dual channel benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
13636XL250Meter with pH/ISE Kit
13636XL250ApH/ISE Meter
13636XL250NMeter with Ammonia Kit
Catalog number 13636XL250
Price (KRW)
-
Product Type:
Meter with pH/ISE Kit
Provide advanced pH and ion measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL250 pH/ISE dual channel benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
  • ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Display two pH or ISE measurements simultaneously with dual channel BNC inputs
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ISE Calibration Points: 2 to 6 points
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: Two BNC, 2 mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Includes:
13-636-XL250A: accumet XL250 pH/ISE dual chan. meter, electrode stand, RS232 and USB cables, 100/240V univ power supply.

13-636-XL250F: accumet XL250 pH/ISE dual chan. meter, fluoride electrode (13-620-629), TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply.

13-636-XL250N: accumet XL250 pH/ISE dual chan. meter, ammonia electrode (13-620-509), TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply.

13-636-XL250: accumet XL250 pH/ISE dual chan. meter, TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply.

XL250ACERT: accumet XL250 pH/ISE dual chan. meter, electrode stand, RS232 and USB cables, 100/240V univ power supply, NIST certif.

XL250CERT: accumet XL250 pH/ISE dual chan. meter, TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply, NIST certif.

Specifications
AccuracyMultiple
Accuracy (ISE)0.5% full scale (monovalent ion), 1% full scale (divalent ion)
Accuracy (mV)±0.2 mV
Accuracy (pH)±0.002
Calibration Points (ISE)2 to 6
Calibration Points (pH)Up to 6 preset or 5 custom
Certifications/ComplianceNIST, ISO, FDA, USDA, EPA
Data Management Details3 to 86400 sec.
Data Points2000
Detection RangeMultiple
Dimensions (L x W x H)24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
Frequency50/60Hz
Height (English)3.89 in.
Height (Metric)9.9 cm
IncludesMeter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply
Inputs2 BNC, 2 ATC
Length (English)9.68 in.
Length (Metric)24.6 cm
ModelXL250 Dual pH/ISE Kit
OutputsRS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Product TypeMeter with pH/ISE Kit
Range (ISE)±2000 mV
ResolutionMultiple
Resolution (mV)0.1mV
Temperature Accuracy±0.3°C
Temperature Resolution0.1°C
Voltage100/240 V
Warranty3 Years
Weight (English)2.5 lb.
Weight (Metric)1.13 kg
Width (Metric)17.3 cm
Channels2
Display TypeLCD
Measurement ModespH, mV, ISE, Ion, Concentration, Temperature
Range (mV)±2000 mV
Range (pH)-2 to 20
Resolution (ISE)2/3/4 Digits
Resolution (pH)0.1/0.01/0.001
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach