Search
| Catalog Number | Volume (Metric) | Graduations | Color | Negative Graduations |
|---|---|---|---|---|
| 1367633F also known as FISHERMIS-1367633F-720/CS | 10 mL | 0.1 mL | Orange | 2 mL |
Designed for the larger-volume user, these pipets are calibrated 'To Deliver' with blow-out, and are packed to permit easy access to several pipets at a time without contaminating the entire case. Similar to Cat. No. 7077, except multi-packed in plastic bags. New ISO color-coding is printed directly on the tear strip for easy identification by size. Reference: ASTM E-714.