Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Samco™ Standard Disposable Transfer Pipettes
Thermo Scientific™

Samco™ Standard Disposable Transfer Pipettes

Single squeeze draws up to 3.2 mL
Have Questions?
Catalog NumberVolume (Metric)Graduated
137119D
also known as FISHERTOPSELLING-137119D-PK
7.5 mLYes
Catalog number 137119D
also known as FISHERTOPSELLING-137119D-PK
Price (KRW)
-
Volume (Metric):
7.5 mL
Graduated:
Yes
Single squeeze draws up to 3.2 mL

Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials.

  • Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel
  • One-piece design molded from see-through, low-density polyethylene
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • In four ready-to-use styles
  • One squeeze draws from 2.0 to 3.3 mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
Specifications
Bulb Draw (Metric)3.2 mL
For Use With (Application)Research
GraduatedYes
Quantity500/Pk.
DisposableSingle-use
Length (Metric)15.5 cm
MaterialPolyethylene
TypeTransfer Pipet
Volume (Metric)7.5 mL
Unit SizePack of 500