Specifications
Accuracy0.6°C Freezing point, 0.2%
CalibrationDistilled Water
Certifications/ComplianceIP65, RoHS, WEEE, CE
Includes2 AAA Batteries
Lighting Type589nm LED
Temperature Compensation20°C
Weight (English)3.5 oz.
Weight (Metric)100 g
TypeHandheld Digital Dual-scale Refractometer
Unit SizeEach