Specifications
Diameter (English) Outer2.5 in.
Diameter (Metric) Outer6.3 cm
For Use With (Application)Collect liquid or light viscous samples from tanks, ponds, lagoons and wells
IncludesCylindrical reservoir chamber, weighted plunger that seals the chamber at the bottom, and cable attachment
Length (English)10 in.
Length (Metric)25 cm
Capacity (Metric)500 mL
MaterialStainless Steel
Product TypeBomb Sampler
Unit SizeCase of 1