96-Well Tall-Chimney PCR Plate
Fisher Scientific™

96-Well Tall-Chimney PCR Plate

Compatible with all leading thermal cyclers and MegaBACE™ capillary sequencers†
Have Questions?
Catalog NumberColor
14230242Natural
Catalog number 14230242
Price (KRW)
-
Color:
Natural
Compatible with all leading thermal cyclers and MegaBACE™ capillary sequencers†

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Highly rigid plate deck
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 340μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumic grid
  • Cleanroom produced

Specifications
For Use With (Application)PCR
Product TypePCR Microplate
RimmedYes
Skirt StyleNon-skirt
Array8 x 12
ColorNatural
FormatStandard
MaterialPolypropylene
No. of Wells96
Volume (Metric) Well340 μL
Unit SizeCase of 50