L-Shaped Cell Spreaders
Fisher Scientific™
L-Shaped Cell Spreaders
Ideal for even spreading of bacterial culture
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
14665231
also known as FIS-14665231
1/Pouch, 100 Pouches/Cs.
1466523010/Pouch, 50 Pouches/Cs.
Catalog number 14665231
also known as FIS-14665231
Price (KRW)
-
Quantity:
1/Pouch, 100 Pouches/Cs.
For spreading of bacterial culture on agar plates
  • Disposable
  • Smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
  • L-shape allows application on the entire Petri dish or plate surface
  • Presterilized blue polypropylene eliminates the need for flaming or autoclaving
Specifications
ColorBlue
DisposableSingle-use
For Use With (Application)Ideal for even spreading of bacterial cultures, smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
For Use With (Equipment)Agar Culture Plate
PackagingIndividually Packaged
Quantity1/Pouch, 100 Pouches/Cs.
ShapeL-Shape
MaterialPolypropylene
Product TypeCell Spreader
SterilitySterile
Unit SizePack of 100