Search
| Catalog Number | Diameter (Metric) | Length (Metric) | Thread Size | Capacity (Metric) |
|---|---|---|---|---|
| 149321D also known as FISHERMIS-149321D-12/PK | 38 mm | 200 mm | 38-430 | 170 mL |
| 14932B also known as FISHERTOPSELLING-14932B-144/PK | 16 mm | 150 mm | 15-415 | 20 mL |
| 14932F also known as FISHERMIS-14932F-48/PK | 25 mm | 200 mm | 24-410 | 70 mL |
Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. Caps are provided with a glued-in, white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations. Pencil marks are easily erased.
For phenolic caps, see Cat. No. 9999.