Specifications
Closure MaterialPTFE
Includes2 clear windows
Quantity2/Pk., 8 Pk./Cs.
Width (Metric) Interior2mm
Capacity (Metric)300 μL
For Use With (Application)Transmission from far ultraviolet through the infrared
For Use With (Equipment)Standard Spectrophotometer Cell Holder
Height (Metric)25 mm
MaterialQuartz
Path Length10 mm
ShapeRectangle
TypeMicro Absorption Cell
Wavelength Range200 to 2500 nm
Width (Metric)12.5 mm
Unit SizePack of 2