General Purpose Ultrasonic Cleaning Solution
Product Image
Fisher Scientific™
General Purpose Ultrasonic Cleaning Solution
Cleans a wide range of contaminants.
Have Questions?
Catalog NumberVolume (English)
1533580
also known as FISHERMIS-1533580-EACH
1 gal.
Catalog number 1533580
also known as FISHERMIS-1533580-EACH
Price (KRW)
-
Volume (English):
1 gal.
Cleans a wide range of contaminants.
  • Specially suited for cleaning plastics, glass, ceramics, rubber, and all types of metals including aluminum and magnesium
  • High-performance, heavy-duty ultrasonic cleaner formulation
  • Fast and long lasting
  • Odorless, ammonia-free and nonflammable with no acid, caustic or fuming problems
  • Safe to handle, mild on hands
  • Dilute in ratios up to 10 parts water to one part solution
  • In 1 gal. (3.8L) bottle
Specifications
AppearanceBlue
CompositionWater 70 to 90%, Pentasodium Triphosphate 10 to 20%, Sodium Bicarbonate 5 to 15%, Sodium Lauryl Sulf
For Use With (Application)For cleaning Plastics, Glass, Ceramics, Rubber and all types of Metals
Volume (English)1 gal.
FormLiquid
Unit SizeEach