150 Homogenizer Package
Fisher Scientific™

150 Homogenizer Package

Fisher Scientific™ 150 Homogenizer Package includes everything needed to process a wide range of laboratory samples, typically in less than 30 seconds. Includes motor, stand assembly and 3 generator probes.
Have Questions?
Catalog NumberVoltage
15340168115 V
Catalog number 15340168
Price (KRW)
8,892,000
Each
Voltage:
115 V
Price (KRW)
8,892,000
Each
Fisher Scientific™ 150 Homogenizer Package includes everything needed to process a wide range of laboratory samples, typically in less than 30 seconds. Includes motor, stand assembly and 3 generator probes.
  • Processes laboratory samples, from 200μL to 100mL
  • Powerful 150w motor
  • Lightweight ergonomic design
  • Quiet operation, under 72db
  • Available with disposable generator probes (adapter for use is included)
  • Cross-contamination can be completely eliminated with the use of plastic generator probes
  • Includes 3 stainless steel generator probes; 5 x 75, 7 x 115, and 10 x 115 mm
Specifications
Diameter (English)2 in.
Diameter (Metric)5.1 cm
For Use With (Application)Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
Height (English)6.5 in.
Height (Metric)16 cm
Includes150 W motor, Flat bottom stainless steel generator probe (5 x 75 mm), Sawtooth stainless steel generator probe (7 x 115 mm), Sawtooth stainless steel generator probe (10 x 115 mm), Stand clamp, Motor stand assembly, Disposable probe
Volume Range (Metric)0.2 to 100 mL
Weight (English)16.4 oz.
Weight (Metric)466 g
Product TypeTissue Homogenizer
Speed5000 to 35,000 rpm
Voltage115 V
Unit SizeEach