Powdered Precision Cleaner
Fisher Scientific™

Powdered Precision Cleaner

Concentrated, anionic detergent for manual and ultrasonic cleaning
Have Questions?
Catalog NumberVolume (English)Container Type
16000104
also known as FISHERTOPSELLING-16000104-EACH
4 lb.Box
Catalog number 16000104
also known as FISHERTOPSELLING-16000104-EACH
Price (KRW)
-
Volume (English):
4 lb.
Container Type:
Box
Concentrated, anionic detergent for manual and ultrasonic cleaning

Alconox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, carbonates.

  • Biodegradable and readily disposable
  • Penetrating wetting power to save time
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing gives reliable results without interfering residues
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100 for typical use (10g in 1L of water)
  • USDA authorized for use in federally inspected meat and poultry plants

Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, pipes, radioactive contaminated articles, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, tanks and reactors, passes inhibitory residue test for water analysis.

Used to remove:
Soil, grit, grime, buffing compound, slime, grease, oils, blood, tissue, salts, deposits, particulates, solvents, chemicals, radioisotopes, radioactive contaminations, silicon oils, mold release agents.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal,stainless steel, porcelain, ceramic, plastic, rubber and fiberglass, can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life 2 years from date of manufacture.

Specifications
AppearanceWhite, Cream
CompositionAnionic Detergent, Sodium Linear Alkylaryl Sulfonate, Alcohol Sulfate, Phosphate, Carbonate, Water
Container TypeBox
For Use With (Application)Cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass
Quantity1 Box or 9 Boxes/Cs.
Volume (English)4 lb.
FormPowder
Product TypePowdered Precision Cleaner
pH9.5
Unit SizeEach
Contents & Storage
Store closed and upright in a cool dry place at 15° to 30°C.