Search
Search

| Catalog Number | Size | Length (English) |
|---|---|---|
| 17100855 also known as FIS-17100855 | 5X-Large | 50 in. |
| 17100849 also known as FIS-17100849 | Small | 35 in. |
| 17100852 also known as FIS-17100852 | X-Large | 41 in. |
| 17100854 also known as FIS-17100854 | 3X-Large | 45 in. |
Note:
Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Medical Blue, Cranberry, Teal, Purple and Blueberry.